Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Small Group Learning: Diffusion and Cell Size
GREENER_CHOICESCI_ED

Catalog No. S280002 Shop All Aldon Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S280002 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S280002 Supplier Innovating Science™ Supplier No. IS3003SGL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Small Group Learning: Diffusion and Cell Size kit allows students to create simulated cells (agar blocks) of different sizes and examine how effectively a substance is able to diffuse into the cell in a set period of time.

  • Why are cells microscopic? The answer relates to the need for the cell to effectively bring materials in and remove waste.
  • In this activity, students will create simulated cells (agar blocks) of different sizes and examine how effectively a substance is able to diffuse into the cell in a set period of time.
  • A special indicator in the cells will allow students to visualize the degree of diffusion.
  • The results will clearly display the fact that a smaller volume creates a more favorable condition for the exchange of material across a cell membrane.
  • Kit contains enough materials for 5 groups of students.
  • Teacher's Manual and Student Study Guide copymasters are included.

Specifications

Certifications/Compliance Next Generation Science Standards (NGSS)
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Life Science
Includes 25 g Agar, 10 mL Bromothymol Blue Concentrate, 25 mL Hydrochloric Acid (2.0M), 6 Agar Block Casting Trays, 5 Plastic Cups, 5 Plastic Knives, 5 Plastic Stirrers
Class Size 5 Lab Groups
Product Type Diffusion and Cell Size Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.