Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smallpox A27L Antibody, Novus Biologicals™
SDP

Catalog No. NB10056486T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056486T 0.025 mg
NB10056486 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056486T Supplier Novus Biologicals Supplier No. NB10056486SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Smallpox A27L Polyclonal specifically detects Smallpox A27L in Virus samples. It is validated for Western Blot.

Specifications

Antigen Smallpox A27L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1-1 ug/ml
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Host Species Rabbit
Immunogen Amino acids 42-56 ([DNEETLKQRLTNLEK) of the A27L protein were used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The Smallpox A27L amino acid sequence used as immunogen for this antibody is 100% homologous in variola virus from human, camel, cow, monkey and rabbit.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.