Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smallpox B5R Antibody (1J2F3), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP224699X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224699X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224699X Supplier Novus Biologicals Supplier No. NBP224699AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Smallpox B5R Monoclonal specifically detects Smallpox B5R in Virus samples. It is validated for Western Blot.

Specifications

Antigen Smallpox B5R
Applications Western Blot
Classification Monoclonal
Clone 1J2F3
Conjugate Alexa Fluor 488
Dilution Western Blot
Gene Accession No. P33823
Gene Alias B5R protein, B6R, B7R, B7R protein, EEV membrane glycoprotein, Glycoprotein, H7R, H7R protein, ORF5R, VARVgp175
Host Species Mouse
Immunogen Amino acids 33-47 (IQEYNLIRTYKKVDK) ofthe B5R protein were used as the immunogen (NCBI Reference Sequence: NP_042218.1)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1486562
Test Specificity The amino acid sequence used as immunogen for this phospho antibody is 100% homologous in Human, Chimpanzee, Dog, Rat, Mouse, Chicken and Zebrafish and 92% homologous in Xenopus.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.