Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smallpox B5R Antibody (1J2F3), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB337319 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337319 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337319 Supplier Novus Biologicals Supplier No. NBP224699MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Smallpox B5R Monoclonal antibody specifically detects Smallpox B5R in Virus samples. It is validated for Western Blot

Specifications

Antigen Smallpox B5R
Applications Western Blot
Classification Monoclonal
Clone 1J2F3
Conjugate mFluor Violet 610 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias B5R protein, B6R, B7R, B7R protein, EEV membrane glycoprotein, Glycoprotein, H7R, H7R protein, ORF5R, VARVgp175
Host Species Mouse
Immunogen Amino acids 33-47 (IQEYNLIRTYKKVDK) ofthe B5R protein were used as the immunogen (NCBI Reference Sequence: NP_042218.1)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1486563
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.