Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMAP Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB332053 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332053 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332053 Supplier Novus Biologicals Supplier No. NBP297382MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMAP Polyclonal antibody specifically detects SMAP in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen SMAP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias small acidic protein, chromosome 11 open reading frame 58, SMAP
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SMAP fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10944
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.