Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCA1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB125843 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB125843 100 μg
NB125844 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB125843 Supplier Novus Biologicals Supplier No. NBP310471100UL

Rabbit Polyclonal Antibody

SMARCA1 Polyclonal specifically detects SMARCA1 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry.

Specifications

Antigen SMARCA1
Applications ChIP Assay, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation, Immunohistochemistry
Formulation PBS buffer, 2% sucrose
Gene Alias ATP-dependent helicase SMARCA1, DKFZp686D1623, EC 3.6.1, EC 3.6.4.-, FLJ41547, global transcription activator homologous sequence, Nucleosome-remodeling factor subunit SNF2L, NURF140, probable global transcription activator SNF2L1, SNF2L, SNF2L1ISWI, SNF2LB, SNF2-like 1, SNF2LT, subfamily a, member 1, sucrose nonfermenting 2-like protein 1, SWI, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin a1, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 1, SWI2
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N terminal region of human SMARCA1 (NP_003060). Peptide sequence EQDTAAVAATVAAADATATIVVIEDEQPGPSTSQEEGAAAAATEATAATE
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6594
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.