Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMARCA1 / SNF2L1 Monoclonal Antibody (2C4), Active Motif™

Rat Monoclonal Antibody
Supplier: Active Motif 61465

Description
Members of this G protein family are characterized by the presence of conserved SNF2 domain, a helicase-like region. They also have an ATPase activity and are thought to have a significant role regulating chromatin remodeling. Although SNF2L1 has a ubiquitous expression pattern, its active form is expressed in neurons suggesting a possible role in promoting neurite outgrowth and regulating neuronal physiology. Principal Names: SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily A member 1 (SMARCA1); SNF2L1; SNF2LB; Possible global transcription activator SNF2L1; SWI2 Official Gene Symbol: SMARCA1 Gen Bank Accession Number: NP_620604 Gene ID: 6594 (Human) Gene Map Locus: Xq25 (Human) SNF2L1, a ∼123KDa protein, is a member of SWI/SNF family of proteins.Specifications
SMARCA1 / SNF2L1 | |
Monoclonal | |
1 mg/mL | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
P28370 | |
Smarca1 | |
This antibody was raised against a recombinant protein corresponding to amino acids 583-784 of human SNF2L. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry, Western Blot | |
2C4 | |
Unconjugated | |
Smarca1 | |
5730494M04Rik; ATP-dependent helicase SMARCA1; BOS_25187; DKFZp686D1623; DNA-dependent ATPase SNF2L; FLJ41547; global transcription activator homologous sequence; hSNF2L; ISWI; Nucleosome-remodeling factor subunit SNF2L; NURF140; Probable global transcription activator SNF2L1; RGD1561046; RP3-353H6.2; Smarca1; Snf2l; SNF2L1; SNF2LB; SNF2-like 1; SNF2LT; sucrose nonfermenting 2-like protein 1; SWI; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 1; SWI2 | |
Rat | |
Protein G | |
RUO | |
6594 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction