Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCA1 / SNF2L1 Monoclonal Antibody (2C4), Active Motif™
SDP

Rat Monoclonal Antibody

$132.55 - $397.65

Specifications

Antigen SMARCA1 / SNF2L1
Clone 2C4
Concentration 1 mg/mL
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3423
SDP
Encompass
View Documents
Active Motif
61466
10 μg
Each for $132.55
Only null left
Add to Cart
 
50-199-3422
SDP
Encompass
View Documents
Active Motif
61465
100 μg
Each for $397.65
Only null left
Add to Cart
 
Description

Description

Members of this G protein family are characterized by the presence of conserved SNF2 domain, a helicase-like region. They also have an ATPase activity and are thought to have a significant role regulating chromatin remodeling. Although SNF2L1 has a ubiquitous expression pattern, its active form is expressed in neurons suggesting a possible role in promoting neurite outgrowth and regulating neuronal physiology. Principal Names: SWI/SNF related matrix associated actin dependent regulator of chromatin subfamily A member 1 (SMARCA1); SNF2L1; SNF2LB; Possible global transcription activator SNF2L1; SWI2 Official Gene Symbol: SMARCA1 Gen Bank Accession Number: NP_620604 Gene ID: 6594 (Human) Gene Map Locus: Xq25 (Human) SNF2L1, a ∼123KDa protein, is a member of SWI/SNF family of proteins.
Specifications

Specifications

SMARCA1 / SNF2L1
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
5730494M04Rik; ATP-dependent helicase SMARCA1; BOS_25187; DKFZp686D1623; DNA-dependent ATPase SNF2L; FLJ41547; global transcription activator homologous sequence; hSNF2L; ISWI; Nucleosome-remodeling factor subunit SNF2L; NURF140; Probable global transcription activator SNF2L1; RGD1561046; RP3-353H6.2; Smarca1; Snf2l; SNF2L1; SNF2LB; SNF2-like 1; SNF2LT; sucrose nonfermenting 2-like protein 1; SWI; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 1; SWI2
Smarca1
IgG2b
Protein G
Antibody
2C4
Immunohistochemistry, Western Blot
Unconjugated
Rat
Human
P28370
6594
This antibody was raised against a recombinant protein corresponding to amino acids 583-784 of human SNF2L.
Primary
-20°C, Avoid Freeze/Thaw Cycles
Smarca1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.