Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCA4 / BRG1 Monoclonal Antibody (3G4), Active Motif™
SDP

Rat Monoclonal Antibody

$156.65 - $620.58

Specifications

Antigen SMARCA4 / BRG1
Clone 3G4
Concentration 1 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-2872
SDP
Encompass
View Documents
Active Motif
39808
10 μg
Each for $156.65
Only null left
Add to Cart
 
50-199-2871
SDP
Encompass
View Documents
Active Motif
39807
100 μg
Each for $620.58
Only null left
Add to Cart
 
Description

Description

Brg1 (Brahma-related gene 1) is an ATPase subunit of SWI2/SNF2-like chromatin-remodeling complexes that enable access of regulatory and effector proteins in transcription, DNA repair and DNA replication. Although Brg1-containing complexes are not essential for general cell survival, they participate in transcriptional regulation of several hundred genes including those involved in interferon and stress response, immune cells differentiation, neurogenesis, cell cycle etc. and is absolutely necessary for mouse embryogenesis. Brg1 is also involved in cell growth arrest, senescence and tumour supression.
Specifications

Specifications

SMARCA4 / BRG1
1 mg/mL
Monoclonal
Liquid
RUO
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
ATP-dependent helicase SMARCA4; b2b508.1Clo; b2b692Clo; BAF190; BAF190A; brahma protein-like 1; Brg1; brg-1; BRG1-associated factor 190A; BRM/SWI2-related gene 1; CSS4; FLJ39786; global transcription activator homologous sequence; homeotic gene regulator; HP1-BP72; hSNF2b; Mitotic growth and transcription activator; MRD16; nuclear protein GRB1; Protein brahma homolog 1; Protein BRG-1; RTPS2; SMARCA4; SNF2; SNF2B; SNF2beta; SNF2-beta; Snf2l4; SNF2LB; SNF2-like 4; sucrose nonfermenting-like 4; SW1/SNF; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 4; SWI2; Transcription activator BRG1
Smarca4
IgG2b
Protein G
Antibody
3G4
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Rat
Human, Mouse
P51532, Q3TKT4
20586, 6597
This SMARCA4 / BRG1 antibody was raised against a recombinant protein corresponding to amino acids 214-279 of mouse BRG1.
Primary
-20°C, Avoid Freeze/Thaw Cycles
Smarca4
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.