Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SMARCA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596029
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA596029 100 μL
1 options

Catalog No. PIPA596029

Supplier: Invitrogen™ PA596029

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELFAHFIQPA AQKTPTSPLK MKPGRPRIKK DEKQNLLSVG DYRHRRTEQE EDEELLTESS KATNVCTRFE DSPSYVKWGK LRDYQVRGLN WLISLYENGI NGILADEMGL GKTLQTISLL GYMKHYRNIP GPHMVLVPKS TLHNWMSEFK RWVPTLRSVC LIGDKEQRAA FVRDVLLPGE WDVCVTSYEM LIKEKSVFKK FNWRYLVIDE AHRIKNEKSK LSEIVREFKT TNRLLLTGTP LQNNLHELWS LLNFLLPDVF NSADDFDSWF DTNNCLGDQK LVERLHMVLR PFLLRRIKAD V; Positive Samples: PC-3, H460, 293T, HepG2, SH-SY5Y, Mouse spleen, Mouse brain, Mouse testis; Cellular Location: Nucleus.

SMARCA5 is a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. SMARCA5 is a component of the chromatin remodeling and spacing factor RSF, a facilitator of the transcription of class II genes by RNA polymerase II. The protein encoded by this gene is a member of the SWI/SNF family of proteins. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The protein encoded by this gene is a component of the chromatin remodeling and spacing factor RSF, a facilitator of the transcription of class II genes by RNA polymerase II. The encoded protein is similar in sequence to the Drosophila ISWI chromatin remodeling protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCA5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SMARCA5
Gene Accession No. O60264, Q91ZW3
Gene Alias 4933427E24Rik; D030040M08Rik; D330027N15Rik; hISWI; hSNF2H; ISWI; MommeD4; mSnf2h; SMARCA5; SNF2H; sucrose nonfermenting protein 2 homolog; sucrose nonfermenting-like 5; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin A5; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A member 5; WCRF135
Gene Symbols SMARCA5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-400 of human SMARCA5/SNF2H (NP_0035923).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8467, 93762
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.