Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCA5/SNF2H Rabbit anti-Human, Mouse, Rat, Clone: S05-9D5, Novus Biologicals™
SDP

Catalog No. NB188537 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188537 100 μg
NB188538 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188537 Supplier Novus Biologicals Supplier No. NBP315093100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SMARCA5/SNF2H Monoclonal antibody specifically detects SMARCA5/SNF2H in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry

Specifications

Antigen SMARCA5/SNF2H
Applications Western Blot, ELISA, Immunofluorescence, Immunocytochemistry
Classification Monoclonal
Clone S05-9D5
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2 μ/mL, Immunocytochemistry 2 μg/mL
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias EC 3.6.1, EC 3.6.4.-, hISWI, hSNF2HSWI/SNF-related matrix-associated actin-dependent regulator of chromatin A5, ISWI, SNF2Hsucrose nonfermenting-like 5, subfamily a, member 5, Sucrose nonfermenting protein 2 homolog, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A member 5, WCRF135
Host Species Rabbit
Immunogen A synthetic peptide of human SMARCA5/SNF2H (Uniprot#: O60264)
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chromatin Modifiers
Primary or Secondary Primary
Gene ID (Entrez) 8467
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.