Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCAL1 / HARP Monoclonal Antibody (4B10), Active Motif™
SDP

Mouse Monoclonal Antibody

$132.55 - $397.65

Specifications

Antigen SMARCAL1 / HARP
Clone 4B10
Concentration 1 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3156
SDP
Encompass
missing translation for 'viewDocuments'
Active Motif
61116
10 μg
Each for $132.55
Only null left
Add to cart
 
50-199-3155
SDP
Encompass
missing translation for 'viewDocuments'
Active Motif
61115
100 μg
Each for $397.65
Only null left
Add to cart
 
Description

Description

PRKCQ is a calcium-independent, novel isoform that is important in transducing signals in T lymphocytes. Interaction of a T lymphocyte with an antigen-presenting cell results in clustering of the T cell antigen receptor (TCR) and the recruitment of a large signaling complex to the site of cell-cell contact. PRKCQ is essential for TCR-mediated activation. PKC theta plays a role in pathways that link the TCR signaling complex to the activation of NF-kappa B in mature T lymphocytes.
Specifications

Specifications

SMARCAL1 / HARP
1 mg/mL
Monoclonal
Liquid
RUO
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
6030401P21Rik; AI851716; ATP-driven annealing helicase; Harp; HepA-related protein; hHARP; mharp; Smarcal1; SMARCA-like protein 1; Sucrose nonfermenting protein 2-like 1; Swi/SNF related matrix associated, actin dependent regulator of chromatin, subfamily a-like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a-like 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A-like protein 1
SMARCAL1
IgG1
Protein G
Antibody
4B10
Immunoprecipitation, Western Blot
Unconjugated
Mouse
Bovine, Human, Mouse
Q8BJL0, Q9NZC9, Q9TTA5
338072, 50485, 54380
This SMARCAL1 / HARP antibody was raised against purifed bovine SMARCAL1 / HARP protein.
Primary
-20°C, Avoid Freeze/Thaw Cycles
SMARCAL1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.