Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCC1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB098432 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB098432 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB098432 Supplier Novus Biologicals Supplier No. NBP298777AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMARCC1 Polyclonal specifically detects SMARCC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SMARCC1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias BAF155SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily C member 1, BRG1-associated factor 155, chromatin remodeling complex BAF155 subunit, CRACC1, mammalian chromatin remodeling complex BRG1-associated factor 155, Rsc8, SRG3, subfamily c, member 1, SWI/SNF complex 155 kDa subunit, SWI/SNF complex subunit SMARCC1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI3
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human SMARCC1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6599
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.