Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCC2/BAF170 Rabbit anti-Human, Biotin, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB057070 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB057070 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB057070 Supplier Novus Biologicals Supplier No. NBP298983B

Rabbit Polyclonal Antibody

SMARCC2/BAF170 Polyclonal specifically detects SMARCC2/BAF170 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SMARCC2/BAF170
Applications Immunofluorescence
Classification Polyclonal
Conjugate Biotin
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias BAF170DKFZp313D0632, BRG1-associated factor 170, chromatin remodeling complex BAF170 subunit, CRACC2, mammalian chromatin remodeling complex BRG1-associated factor 170, Rsc8, subfamily c, member 2, SWI/SNF complex 170 kDa subunit, SWI/SNF complex subunit SMARCC2, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily C member 2, SWI3-like protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human SMARCC2/BAF170.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6601
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.