Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCD1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB054073 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB054073 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB054073 Supplier Novus Biologicals Supplier No. NBP298471AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMARCD1 Polyclonal specifically detects SMARCD1 in Human samples. It is validated for Western Blot.

Specifications

Antigen SMARCD1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias BAF60Amammalian chromatin remodeling complex BRG1-associated factor 60A, BRG1-associated factor 60A, chromatin remodeling complex BAF60A subunit, CRACD1, Rsc6p, subfamily d, member 1, SWI/SNF complex 60 kDa subunit, SWI/SNF complex 60 kDa subunit A, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily D member 1,60 kDa BRG-1/Brm-associated factor subunit A, Swp73-like protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SMARCD1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 6602
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.