Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCD1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP188719 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP188719 0.1 mL
NB431161 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP188719 Supplier Novus Biologicals Supplier No. NBP188719
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

SMARCD1 Polyclonal specifically detects SMARCD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SMARCD1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias BAF60Amammalian chromatin remodeling complex BRG1-associated factor 60A, BRG1-associated factor 60A, chromatin remodeling complex BAF60A subunit, CRACD1, Rsc6p, subfamily d, member 1, SWI/SNF complex 60 kDa subunit, SWI/SNF complex 60 kDa subunit A, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily D member 1,60 kDa BRG-1/Brm-associated factor subunit A, Swp73-like protein
Gene Symbols SMARCD1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:MGPAPGQGLYRSPMPGAAYPRPGMLPGSRMTPQGPSMGPPGYGGNPSVRPGLAQSGMDQSRKRPAPQQIQQVQQQAVQNRNHNAKK
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 6602
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.