Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SMART Digest™ Trypsin Kits

Catalog No. 03251893
Click to view available options
Format:
Bulk Low pH Magnetic Resin
Bulk Low pH Resin
Bulk Magnetic Resin
Bulk Non-magnetic Resin
Resin in Pre-filled PCR Tubes
Soluble Trypsin
Type:
Soluble Trypsin Kit
Soluble Trypsin Kit - Low Salt
Soluble Trypsin Kit - Low pH
Trypsin Kit
Includes:
Low pH Resin, Collection Plate
Resin, Buffer
Resin, Buffer, Collection Plate, Filter Plate
Resin, Buffer, Collection Plate, SOLAμ SPE Plate
Resin, Collection Plate
Resin, Collection Plate, Filter Plate
Resin, Collection Plate, SOLAμ SPE Plate
Soluble Trypsin, Buffer, Collection Plate
Soluble Trypsin, Low pH Buffer, Collection Plate

Catalog No. 03-251-893

Supplier: Thermo Scientific™ 60109101BLPH

Only null left
Add to Cart
Add to Cart

Obtain fast, simple, and highly reproducible trypsin protein digestion for peptide characterization and quantitation in biopharmaceutical protein research.

Obtain high quality analytical results from protein digests using Thermo Scientific™ SMART Digest™ Kits. A significant advance in sample preparation for biopharmaceutical protein research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. The SMART Digest Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.

Trypsin Protein Digestion

SMART Digest kits enable you to characterize and quantitate proteins faster, easier, and quicker than ever before. This version of the kit uses trypsin, which cleaves peptides on the C-terminal side of lysine and arginine amino acid residues.

Easy and simple
  • Significantly increased speed of sample processing
  • Simple process
  • Easily automate entire protocol for high-throughput processing
Reliable Results
  • Increased reproducibility over existing protocols
  • Higher sensitivity
  • Higher data confidence
Significantly Reduces Sample Preparation Time

Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours.

Cleaner Samples

Reduction in the number of chemicals used and the immobilized trypsin design helps mitigate against chemically induced post translational modifications (PTMs) and autolysis. This results in results which are easier to interpret.

Couple with SOLAμ SPE Plates for Desalting

When used with the SMART Digest kits for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.

TRUSTED_SUSTAINABILITY

Specifications

Type Trypsin Kit
Description Smart digest kit with bulk resin , low pH
Format Bulk Low pH Resin
Enzyme Trypsin
Cleavage C-terminal side of lysine and arginine amino acids
Capture None
Product Line SMART Digest
For Use With (Application) MAM
Includes Resin, Collection Plate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.