Learn More
Thermo Scientific™ Smart-Vue™ Event Notification Hardware
Receive immediate notification by means of your preferred alert method, including audible/visual siren, fax, telephone, email or text messaging.
$1145.23 - $1731.10
Specifications
Certifications/Compliance Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
For Use With (Application) Cold Storage
For Use With (Equipment) Standard RJ11 Telephone Line
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
SV500500LSB
View Documents
Thermo Scientific™
SV500500LSB
Smart-Vue Audible and Visual Alarm, 915MHz
Each for $1,731.10
Only null left
Add to Cart
SV509500LSB
View Documents
Thermo Scientific™
SV509500LSB
Smart-Vue Voice and Fax Alert
Each for $1,145.23
Only null left
Add to Cart
Description
Description
Receive immediate notification by means of your preferred alert method, including audible/visual siren, fax, telephone, email or text messaging. Thermo Scientific™ Smart-Vue™ Event Notification Hardware delivers the continuous monitoring, secure data logging and automatic audit trails required to comply with SOP, cGxP and 21 CFR part 11 regulatory standards.
Local RF Audible and Visual Alarm:
  • Universal (100V.240V) to 12 volt power supply
  • 3.6V battery
  • Velcro
  • Hardware
  • Customer reference letter
Voice and Fax Alert:
  • USB 1.0 or 2.0 powered
  • Fits standard RJ11 telephone line
Ordering Information:
Availability of Smart-Vue products vary by country. Please ask your local Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.
Specifications
Specifications
Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Cold Storage
Standard RJ11 Telephone Line
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.