Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SMBA 1
GREENER_CHOICE

Catalog No. p-5546190
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity and selective activator of Bax

SMBA 1 is a high affinity and selective activator of Bax (Ki = 43.3 nM). Binds to the S184 binding pocket to block Bax phosphorylation. Does not bind to other Bcl-2 family members, including Bcl-2, Bak and Bid. Facilitates Bax insertion into the mitochondrial membrane and promotes Bax-dependent cytochrome c release. Induces apoptosis in lung cancer cell lines expressing high levels of Bax selectively over normal cells expressing low levels of Bax. Represses tumor growth of A549 lung cancer xenografts in mice.

Specifications

Quantity 10 mg
Product Type SMBA 1
Purity >98%
Molecular Weight (g/mol) 315.32
Inhibitors SMBA 1

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.