Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC1 Antibody (8C11), Novus Biologicals™
SDP

Catalog No. NB298350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298350 50 μL
NB298351 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298350 Supplier Novus Biologicals Supplier No. NBP32631350UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SMC1 Monoclonal antibody specifically detects SMC1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen SMC1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 8C11
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias CDLS2, DXS423ESMC1 structural maintenance of chromosomes 1-like 1 (yeast), KIAA0178DKFZp686L19178, Sb1.8, SB1.8MGC138332, segregation of mitotic chromosomes 1, SMC protein 1A, SMC1 (structural maintenance of chromosomes 1, yeast)-like 1, SMC1 structural maintenance of chromosomes 1-like 1, SMC-1A, SMC-1-alpha, SMC1L1, SMC1SMC1alpha, SMCB, structural maintenance of chromosomes 1A, structural maintenance of chromosomes protein 1A
Host Species Rabbit
Immunogen A synthesized peptide derived from Human SMC1 [UniProt Q14683]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 8243
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.