Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB289949 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289949 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289949 Supplier Novus Biologicals Supplier No. NBP232142PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMC1 Polyclonal antibody specifically detects SMC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation

Specifications

Antigen SMC1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation
Formulation PBS
Gene Alias CDLS2, DXS423ESMC1 structural maintenance of chromosomes 1-like 1 (yeast), KIAA0178DKFZp686L19178, Sb1.8, SB1.8MGC138332, segregation of mitotic chromosomes 1, SMC protein 1A, SMC1 (structural maintenance of chromosomes 1, yeast)-like 1, SMC1 structural maintenance of chromosomes 1-like 1, SMC-1A, SMC-1-alpha, SMC1L1, SMC1SMC1alpha, SMCB, structural maintenance of chromosomes 1A, structural maintenance of chromosomes protein 1A
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region of human SMC1 (structural maintenance of chromosomes 1-like 1 ) surrounding serine 957 using the numbering given in entry NP_006297.2 (Gene ID 8243) when the serine residue is phosphorylated.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 8243
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.