Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB100373646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100373646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100373646 Supplier Novus Biologicals Supplier No. NB100373JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMC2 Polyclonal specifically detects SMC2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, KnockDown, Western Blot.

Specifications

Antigen SMC2
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, KnockDown, Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CAP-E, CAPESMC2 structural maintenance of chromosomes 2-like 1 (yeast), Chromosome-associated protein E, FLJ10093, hCAP-EXCAP-E homolog, SMC protein 2, SMC-2, SMC2 (structural maintenance of chromosomes 2, yeast)-like 1, SMC2 structural maintenance of chromosomes 2-like 1, SMC2L1, structural maintenance of chromosomes (SMC) family member, structural maintenance of chromosomes 2, structural maintenance of chromosomes protein 2
Gene Symbols SMC2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1150 and the C-terminus (residue 1197) of human Structural Maintenance of Chromosomes 2 using the numbering given in entry NP_006435.1 (GeneID 10592).
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10592
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.