Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC2 Antibody, Novus Biologicals™
SDP

Catalog No. NB100373 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100373 Supplier Novus Biologicals Supplier No. NB100373
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

SMC2 Polyclonal specifically detects SMC2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, KnockDown, Western Blot.

Specifications

Antigen SMC2
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, KnockDown, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:400, Immunoprecipitation 2-10 ug/mg lysate, Knockdown Validated
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_006435.1
Gene Alias CAP-E, CAPESMC2 structural maintenance of chromosomes 2-like 1 (yeast), Chromosome-associated protein E, FLJ10093, hCAP-EXCAP-E homolog, SMC protein 2, SMC-2, SMC2 (structural maintenance of chromosomes 2, yeast)-like 1, SMC2 structural maintenance of chromosomes 2-like 1, SMC2L1, structural maintenance of chromosomes (SMC) family member, structural maintenance of chromosomes 2, structural maintenance of chromosomes protein 2
Gene Symbols SMC2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1150 and the C-terminus (residue 1197) of human Structural Maintenance of Chromosomes 2 using the numbering given in entry NP_006435.1 (GeneID 10592).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10592
Test Specificity Human. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Crab-eating macaque and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.