Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC3 Rabbit anti-Human, Mouse, Rat, Clone: 8W9R7, Novus Biologicals™
SDP

Catalog No. NB166691 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166691 100 μg
NB166692 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166691 Supplier Novus Biologicals Supplier No. NBP315798100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SMC3 Monoclonal antibody specifically detects SMC3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation

Specifications

Antigen SMC3
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 8W9R7
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Bamacan, BAMSMC protein 3, Basement membrane-associated chondroitin proteoglycan, BMH, Chondroitin sulfate proteoglycan 6, chondroitin sulfate proteoglycan 6 (bamacan), Chromosome-associated polypeptide, CSPG6structural maintenance of chromosomes protein 3, hCAP, HCAPbamacan, SMC-3, SMC3L1CDLS3, structural maintenance of chromosomes 3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1118-1217 of human SMC3 (Q9UQE7).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 9126
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.