Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC4 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB336343 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB336343 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB336343 Supplier Novus Biologicals Supplier No. NBP297937APC

Rabbit Polyclonal Antibody

SMC4 Polyclonal antibody specifically detects SMC4 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen SMC4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias CAP-C, CAPCSMC4 structural maintenance of chromosomes 4-like 1 (yeast), Chromosome-associated polypeptide C, hCAP-CSMC4 (structural maintenance of chromosomes 4, yeast)-like 1, SMC protein 4, SMC-4, SMC4 structural maintenance of chromosomes 4-like 1, SMC4L1, structural maintenance of chromosomes 4, structural maintenance of chromosomes protein 4, XCAP-C homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SMC4 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10051
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.