Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC4 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB049659 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB049659 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB049659 Supplier Novus Biologicals Supplier No. NBP297937R

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

SMC4 Polyclonal specifically detects SMC4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SMC4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CAP-C, CAPCSMC4 structural maintenance of chromosomes 4-like 1 (yeast), Chromosome-associated polypeptide C, hCAP-CSMC4 (structural maintenance of chromosomes 4, yeast)-like 1, SMC protein 4, SMC-4, SMC4 structural maintenance of chromosomes 4-like 1, SMC4L1, structural maintenance of chromosomes 4, structural maintenance of chromosomes protein 4, XCAP-C homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SMC4 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10051
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.