Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC4 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056394 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056394 0.1 mg
NB10056394T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056394 Supplier Novus Biologicals Supplier No. NB10056394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMC4 Polyclonal specifically detects SMC4 in Human samples. It is validated for Flow Cytometry, Western Blot.

Specifications

Antigen SMC4
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Flow Cytometry
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. NP_001099000
Gene Alias CAP-C, CAPCSMC4 structural maintenance of chromosomes 4-like 1 (yeast), Chromosome-associated polypeptide C, hCAP-CSMC4 (structural maintenance of chromosomes 4, yeast)-like 1, SMC protein 4, SMC-4, SMC4 structural maintenance of chromosomes 4-like 1, SMC4L1, structural maintenance of chromosomes 4, structural maintenance of chromosomes protein 4, XCAP-C homolog
Gene Symbols SMC4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 281-297 of human CAP-C. Genbank accession no. NP_001002800.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10051
Test Specificity Cross reacts with Human. Predicted to react with Mouse, Rat, Xenopus and Chicken.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.