Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC4 Rabbit anti-Human, Mouse, Rat, Clone: 9Z8S3, Novus Biologicals™
SDP

Catalog No. NB167529 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167529 100 μg
NB167530 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167529 Supplier Novus Biologicals Supplier No. NBP316219100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SMC4 Monoclonal antibody specifically detects SMC4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen SMC4
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 9Z8S3
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CAP-C, CAPCSMC4 structural maintenance of chromosomes 4-like 1 (yeast), Chromosome-associated polypeptide C, hCAP-CSMC4 (structural maintenance of chromosomes 4, yeast)-like 1, SMC protein 4, SMC-4, SMC4 structural maintenance of chromosomes 4-like 1, SMC4L1, structural maintenance of chromosomes 4, structural maintenance of chromosomes protein 4, XCAP-C homolog
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1189-1288 of human SMC4 (Q9NTJ3).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10051
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.