Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC5 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB10469A594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10469A594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10469A594 Supplier Novus Biologicals Supplier No. NB100469AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMC5 Polyclonal specifically detects SMC5 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.

Specifications

Antigen SMC5
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias hSMC5, KIAA0594SMC5 structural maintenance of chromosomes 5-like 1 (yeast), SMC protein 5, SMC-5, SMC5L1SMC5 structural maintenance of chromosomes 5-like 1, structural maintenance of chromosomes 5, structural maintenance of chromosomes protein 5
Gene Symbols SMC5
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 1050 and the C-terminus (residue 1101) of human Structural Maintenance of Chromosomes 5 using the numbering given in TrEMBL entry Q8IY18 (GeneID 23137).
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 23137
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.