Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMCO3 Antibody, Novus Biologicals™
SDP

Catalog No. NB437557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB437557 25ul
NBP230513 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB437557 Supplier Novus Biologicals Supplier No. NBP23051325UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMCO3 Polyclonal specifically detects SMCO3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen C12orf69
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. A2RU48
Gene Alias Chromosome 12 Open Reading Frame 69, LOC440087, Uncharacterized Protein C12orf69
Gene Symbols C12orf69
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: MAQSDFLYPENPKRREEVNRLHQQLLDCLSDSFDVTNKLTEVLNMHLGRRLASIEMKRDGTIKENCDLIIQAIMK
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440087
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.