Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMG5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00
Specifications
Antigen | SMG5 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15677820
![]() |
Novus Biologicals
NBP15677820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP156778
![]() |
Novus Biologicals
NBP156778 |
100 μL | N/A | N/A | N/A | ||||
Description
SMG5 Polyclonal specifically detects SMG5 in Human samples. It is validated for Western Blot.Specifications
SMG5 | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication | |
EST1BFLJ12287, EST1-like protein B, Est1p-like protein B, ever shorter telomeres 1B, hSMG-5, KIAA1089FLJ34864, LPTS interacting protein, LPTS-interacting protein, LPTSRP1, LPTS-RP1EST1 telomerase component homolog B, protein SMG5, RP11-54H19.7, SMG-5, SMG-5 homolog, Smg-5 homolog, nonsense mediated mRNA decay factor (C. elegans) | |
SMG5 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Q9UPR3 | |
23381 | |
Synthetic peptides corresponding to SMG5(Smg-5 homolog, nonsense mediated mRNA decay factor (C. elegans)) The peptide sequence was selected from the N terminal of SMG5. Peptide sequence MSQGPPTGESSEPEAKVLHTKRLYRAVVEAVHRLDLILCNKTAYQEVFKP. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title