Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMG5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP147272 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147272 Supplier Novus Biologicals Supplier No. NBP147272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMG5 Polyclonal specifically detects SMG5 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen SMG5
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2-5 μg/mg lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Alias EST1BFLJ12287, EST1-like protein B, Est1p-like protein B, ever shorter telomeres 1B, hSMG-5, KIAA1089FLJ34864, LPTS interacting protein, LPTS-interacting protein, LPTSRP1, LPTS-RP1EST1 telomerase component homolog B, protein SMG5, RP11-54H19.7, SMG-5, SMG-5 homolog, Smg-5 homolog, nonsense mediated mRNA decay factor (C. elegans)
Gene Symbols SMG5
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 400 and 450 of human Suppressor with Morphogenetic Effects on Genitalia 5 using the numbering given in entry NP_056142.2 (GeneID 23381).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23381
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.