Learn More
Description
Specifications
Specifications
| Antigen | SMG6 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | Q86US8 |
| Gene Alias | C17orf31, EST1 telomerase component homolog A, EST1A, EST1-like protein A, ever shorter telomeres 1A, hSmg5/7a, KIAA0732, SMG-6, smg-6 homolog, nonsense mediated mRNA decay factor (C. elegans), telomerase subunit EST1A, telomerase-binding protein EST1A |
| Gene Symbols | SMG6 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
