Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMN Antibody (2B1), Novus Biologicals™
SDP

Catalog No. NB1001936 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001936 0.1 mL
NB1001936SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1001936 Supplier Novus Biologicals Supplier No. NB1001936
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 21 publication

SMN Monoclonal specifically detects SMN in Human, Mouse, Primate, Rat, Xenopus species, Xenopus species (African Clawed Frog), Zebrafish samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen SMN
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2B1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 30102724), ELISA reported in scientific literature (PMID 23973875), Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:250, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200
Gene Accession No. Q16637, P97801, O35876
Gene Alias Gemin-1, Kugelberg-Welander disease), SMNC, survival motor neuron protein, survival of motor neuron 1, telomeric
Gene Symbols SMN1
Host Species Mouse
Immunogen Purified recombinant His6-tagged human SMN protein. [Swiss-Prot# Q16637]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Neurodegeneration, Neuroscience, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6606
Test Specificity Human, mouse, rat, monkey and Xenopus. Other species not tested.
Target Species Human, Mouse, Mouse, Primate, Rat, Xenopus species, Xenopus species, Zebrafish
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.