Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMN Antibody (2B1), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB1001936X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001936X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001936X Supplier Novus Biologicals Supplier No. NB1001936AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SMN Monoclonal specifically detects SMN in Human, Mouse, Primate, Rat, Xenopus species, Zebrafish samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen SMN
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2B1
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Accession No. Q16637
Gene Alias Gemin-1, Kugelberg-Welander disease), SMNC, survival motor neuron protein, survival of motor neuron 1, telomeric
Gene Symbols SMN1
Host Species Mouse
Immunogen Purified recombinant His6-tagged human SMN protein. [Swiss-Prot# Q16637]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Neurodegeneration, Neuroscience, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6606
Test Specificity Human, mouse, rat, monkey and Xenopus. Other species not tested.
Target Species Human, Mouse, Primate, Rat, Xenopus species, Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.