Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMN Mouse anti-Human, Mouse, Rat, Clone: 4B7, Novus Biologicals™
SDP

Catalog No. NB151825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151825 100 μg
NB151826 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151825 Supplier Novus Biologicals Supplier No. NBP311944100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SMN Monoclonal antibody specifically detects SMN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen SMN
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 4B7
Conjugate Unconjugated
Dilution Western Blot 1:4000, Immunocytochemistry/Immunofluorescence 1:50-1:500
Formulation 100 ul in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol.
Gene Alias Gemin-1, Kugelberg-Welander disease), SMNC, survival motor neuron protein, survival of motor neuron 1, telomeric
Host Species Mouse
Immunogen Recombinant human his-tagged SMN protein expressed in and purified from E. coli. The epitope recognized by the 4B7 clone binds to the region encoded by exon 1. (UniProt: Q16637)
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Neurodegeneration, Neuroscience, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6606
Target Species Human, Mouse, Rat
Content And Storage Store at -20C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.