Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMNDC1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB291418 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291418 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291418 Supplier Novus Biologicals Supplier No. NBP147302APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMNDC1 Polyclonal antibody specifically detects SMNDC1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen SMNDC1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias SMNRSMN-related protein, SPF30splicing factor 30, survival of motor neuron-related, survival motor neuron domain containing 1,30 kDa splicing factor SMNrp, Survival motor neuron domain-containing protein 1, survival of motor neuron-related-splicing factor 30
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 188 and 238 of human Splicing Factor 30 using the numbering given in entry NP_005862.1 (GeneID 10285).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10285
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.