Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMNDC1 Antibody (PCRP-SMNDC1-1A9), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB382586 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382586 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382586 Supplier Novus Biologicals Supplier No. NBP308232FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SMNDC1 Monoclonal antibody specifically detects SMNDC1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen SMNDC1
Applications Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-SMNDC1-1A9
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias SMNRSMN-related protein, SPF30splicing factor 30, survival of motor neuron-related, survival motor neuron domain containing 1,30 kDa splicing factor SMNrp, Survival motor neuron domain-containing protein 1, survival of motor neuron-related-splicing factor 30
Host Species Mouse
Immunogen Recombinant full-length human SMNDC1 protein (Uniprot: O75940 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10285
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.