Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMOC-2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP139788 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP139788 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP139788 Supplier Novus Biologicals Supplier No. NBP139788
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMOC-2 Polyclonal specifically detects SMOC-2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SMOC-2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA 1 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 20 ug/ml
Formulation PBS and 50% Glycerol with 0.01% Sodium Azide
Gene Alias bA270C4A.1, bA37D8.1, dJ421D16.1, MST117, MSTP117, MSTP140, SMAP-2, SMAP2Secreted modular calcium-binding protein 2, SMOC-2, smooth muscle associated protein 2, Smooth muscle-associated protein 2, SPARC related modular calcium binding 2, SPARC-related modular calcium-binding protein 2, thyroglobulin type-1 repeat containing protein
Gene Symbols SMOC2
Host Species Rabbit
Immunogen Synthetic peptide from human SMOC-2.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64094
Test Specificity Synthetic peptide corresponding to N-terminal residues of human SMOC (Secreted modular calcium-binding protein 2) or SPARC related modular calcium-binding protein 2 precursor, Smooth muscle-associated.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.