Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smoothelin Antibody (885424), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB302576 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302576 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302576 Supplier Novus Biologicals Supplier No. FAB8278N

Mouse Monoclonal Antibody

Smoothelin Monoclonal antibody specifically detects Smoothelin in Human samples. It is validated for Immunohistochemistry, Flow Cytometry, CyTOF

Specifications

Antigen Smoothelin
Applications Immunohistochemistry, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 885424
Conjugate Alexa Fluor 700
Dilution Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FLJ35365, FLJ38597, smoothelin, SMSMO
Host Species Mouse
Immunogen E. coli-derived recombinant human Smoothelin B, Met1-Leu132, Accession # P53814
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6525
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.