Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smopex™-111, metal scavenger, Thermo Scientific™

Catalog No. AA4502714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4502714 25 g
AA4502706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4502714 Supplier Thermo Scientific Chemicals Supplier No. 4502714

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Recovery of precious metals from catalyst processes, especially Pd, Rh, Ru, Pt, and Ir
  • Also, heavy metals removed: Cu and Hg
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536755-37-0
Synonym Styryl thiol grafted polyethylene fiber

Specifications

Color White
Physical Form Powder
Density 0.3g/mL
Quantity 25 g
Chemical Name or Material Smopex™-111 metal scavenger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.