Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMYD2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB291236 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291236 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291236 Supplier Novus Biologicals Supplier No. NBP150043F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SMYD2 Polyclonal antibody specifically detects SMYD2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen SMYD2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias EC 2.1.1.43, Histone methyltransferase SMYD2, HSKM-Bzinc finger, MYND domain containing 14, KMT3CEC 2.1.1.-, Lysine N-methyltransferase 3C, MGC119305, SET and MYND domain containing 2, SET and MYND domain-containing protein 2, ZMYND14
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 383 and 433 of human SET and MYND Domain Containing 2 using the numbering given in entry NP_064582.2 (GeneID 56950).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56950
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.