Learn More
SNAI1 / Snail Monoclonal Antibody (9H2), Active Motif™

Description
Specifications
Specifications
| Antigen | SNAI1 / Snail |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 9H2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene | Snai1 |
| Gene Accession No. | O95863 |
| Gene Alias | AI194338; dJ710H13.1; Protein sna; Protein snail homolog 1; SLUGH2; Sna; Sna protein; Sna1; SNAH; Snai1; SNAIL; snail 1; snail 1 homolog; snail 1 zinc finger protein; snail 1, zinc finger protein; snail family transcriptional repressor 1; snail family zinc finger 1; snail homolog 1; Snail1; Zinc finger protein SNAI1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.