Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNAI1 / Snail Monoclonal Antibody (9H2), Active Motif™

Rat Monoclonal Antibody
$135.30 - $405.90
Specifications
| Antigen | SNAI1 / Snail |
|---|---|
| Clone | 9H2 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3351
![]()
|
Active Motif
61368 |
10 μg |
Each for $135.30
|
|
|||||
50-199-3350
![]()
|
Active Motif
61367 |
100 μg |
Each for $405.90
|
|
|||||
Description
Zinc finger protein SNAI1 (SNAIL)is a zinc finger transcriptional repressor which down regulates the expression of the ectodermal gene within the mesoderm and functions to form the mesoderm and neural crest. It is reported that SNAI1 acts as an important factor of tumor invasion as evidenced by its role in E-cadherin down-regulation and induction of epithelial-mesenchymal transition (EMT). In human breast cancer, the expression of SNAI1 and/or the homologous SNAI2 (Slug) has been associated with E-cadherin repression, local or distant metastasis, tumor recurrence, or poor prognosis in different tumor series. It is also reported that Snail 1 protein in the stromamay be anew putative prognosis marker for colon tumors.Specifications
| SNAI1 / Snail | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
| AI194338; dJ710H13.1; Protein sna; Protein snail homolog 1; SLUGH2; Sna; Sna protein; Sna1; SNAH; Snai1; SNAIL; snail 1; snail 1 homolog; snail 1 zinc finger protein; snail 1, zinc finger protein; snail family transcriptional repressor 1; snail family zinc finger 1; snail homolog 1; Snail1; Zinc finger protein SNAI1 | |
| Snai1 | |
| IgG2a | |
| Protein G | |
| Antibody |
| 9H2 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rat | |
| Human | |
| O95863 | |
| 6615 | |
| This antibody was raised against a full-length recombinant protein corresponding to human SNAI1. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Snai1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title