Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Snail Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP227293 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227293 0.1 mg
NBP227293SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227293 Supplier Novus Biologicals Supplier No. NBP227293
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

Snail Polyclonal specifically detects Snail in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Snail
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml. Use reported in scientific literature (PMID 28260923), Simple Western 1:500, Immunohistochemistry 1:200. Use reported in scientific literature (PMID 30488343), Immunohistochemistry-Paraffin 1:200
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. O95863
Gene Alias Protein sna, Protein snail homolog 1, SLUGH2, SNA, SNAHdJ710H13.1, snail 1 (drosophila homolog), zinc finger protein, snail 1 homolog, snail 1 zinc finger protein, snail 1, zinc finger protein, snail homolog 1 (Drosophila), zinc finger protein SNAI1
Gene Symbols SNAI1
Host Species Rabbit
Immunogen A portion of amino acids 1-60 of human SNAI1 was used as the immunogen.
Purification Method Immunogen affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Developmental Biology, Hypoxia, Lipid and Metabolism, Metastasis, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6615
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 94% homologous in mouse and rat. It is 61% homologous with human SNAI2 protein.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.