Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Snail Antibody, Novus Biologicals™
SDP

Catalog No. NBP227184SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227184SS 0.025 mg
NBP227184 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227184SS Supplier Novus Biologicals Supplier No. NBP227184SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Snail Polyclonal specifically detects Snail in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.

Specifications

Antigen Snail
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoassay
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3-5ug/ml, Immunocytochemistry/Immunofluorescence 1:100, Knockout Validated reported in scientific literature (PMID 32180899)
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. O95863
Gene Alias Protein sna, Protein snail homolog 1, SLUGH2, SNA, SNAHdJ710H13.1, snail 1 (drosophila homolog), zinc finger protein, snail 1 homolog, snail 1 zinc finger protein, snail 1, zinc finger protein, snail homolog 1 (Drosophila), zinc finger protein SNAI1
Gene Symbols SNAI1
Host Species Rabbit
Immunogen A portion of amino acids 80-130 of human SNAI1/Snail was used as the immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Developmental Biology, Hypoxia, Lipid and Metabolism, Metastasis, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6615
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, horse and dog, and 94% homologous mouse and rat.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.