Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Snail, p Ser246 Antibody, Novus Biologicals™
SDP

Catalog No. NBP25477605 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25477605 0.05 mg
NBP254776 0.1 mg
NB413812 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP25477605 Supplier Novus Biologicals Supplier No. NBP2547760.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Snail Polyclonal specifically detects Snail in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence.

Specifications

Antigen Snail
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunofluorescence 1:100-1:200
Formulation Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl and 50% glycerol. with 0.02% sodium azide
Gene Alias Protein sna, Protein snail homolog 1, SLUGH2, SNA, SNAHdJ710H13.1, snail 1 (drosophila homolog), zinc finger protein, snail 1 homolog, snail 1 zinc finger protein, snail 1, zinc finger protein, snail homolog 1 (Drosophila), zinc finger protein SNAI1
Gene Symbols SNAI1
Host Species Rabbit
Immunogen SNAI1 (phospho Ser246) antibody was raised against a peptide sequence around phosphorylation site of Serine 246 (T-F-S (p)-R-M) derived from Human SNAI1.
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 6615
Test Specificity The antibody detects endogenous levels of SNAI1 only when phosphorylated at serine 246.
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.