Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Snail, p Ser246 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $706.00
Specifications
Concentration | 1 mg/mL |
---|---|
Dilution | Western Blot 1:500-1:1000, Immunofluorescence 1:100-1:200 |
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP254776
![]() |
Novus Biologicals
NBP254776 |
0.1 mg |
Each for $706.00
|
|
|||||
NBP25477605
![]() |
Novus Biologicals
NBP2547760.05MG |
0.05 mg |
Each for $450.00
|
|
|||||
NB413812
![]() |
Novus Biologicals
NBP2547760.025MG |
0.025 mg |
Each for $196.10
|
|
|||||
Description
Snail Polyclonal specifically detects Snail in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence.Specifications
1 mg/mL | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Human, Mouse | |
6615 | |
SNAI1 (phospho Ser246) antibody was raised against a peptide sequence around phosphorylation site of Serine 246 (T-F-S (p)-R-M) derived from Human SNAI1. | |
Primary |
Western Blot 1:500-1:1000, Immunofluorescence 1:100-1:200 | |
Polyclonal | |
Rabbit | |
Protein sna, Protein snail homolog 1, SLUGH2, SNA, SNAHdJ710H13.1, snail 1 (drosophila homolog), zinc finger protein, snail 1 homolog, snail 1 zinc finger protein, snail 1, zinc finger protein, snail homolog 1 (Drosophila), zinc finger protein SNAI1 | |
SNAI1 | |
IgG | |
The antibody detects endogenous levels of SNAI1 only when phosphorylated at serine 246. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title