Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SNAP 94847 hydrochloride
GREENER_CHOICE

Catalog No. 334710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-471-0 10 mg
33-475-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-471-0 Supplier Tocris Bioscience™ Supplier No. 3347/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective MCH1 antagonist

Potent melanin-concentrating hormone receptor 1 (MCH1) antagonist (Ki = 2.2 nM, KD = 530 pM) that displays > 80-fold and > 500-fold selectivity over α1A and D2 receptors respectively. Increases neurogenesis in the dentate gyrus and decreases food-reinforced operant responding in vivo. Exhibits anxiolytic activity and is orally active.

Chemical Identifiers

CAS 487051-12-7
Molecular Formula C29H33ClF2N2O2

Specifications

Quantity 10 mg
Formula Weight 515.03
Purity >98%
Molecular Weight (g/mol) 515.042
Chemical Name or Material SNAP 94847 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.