Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAP tag Antibody, Novus Biologicals™
SDP

Catalog No. NBP237817 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237817 100 μg
NB397071 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP237817 Supplier Novus Biologicals Supplier No. NBP237817
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SNAP tag Polyclonal specifically detects SNAP tag in Non-species specific samples. It is validated for Western Blot.

Specifications

Antigen SNAP tag
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml
Formulation BBS (pH 8.4) with No Preservative
Host Species Rabbit
Immunogen purified recombinant SNAP
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects SNAP in samples. This product has been successfully used in Western Blot applications. The immunogen is purified recombinant SNAP.
Target Species Non-species specific
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.